Learn More
Medchemexpress LLC Vancomycin (Standard) | 1404-90-6 | 95.92% | 50 MG
SDP
Catalog No. 5000408560
Encompass_Preferred
Vancomycin (Standard) is the analytical standard of Vancomycin, intended for research and analytical applications. It is an antibiotic used for the treatment of bacterial infections.
  • The compound is the grade of analytical standard, which is the reference standard supplied assay.
  • It is commonly used in qualitative, quantitative, and methodological research experiments in HPLC, GC, and MS.
  • Application in disease research: Antibacterial.
  • Initial source: Endogenous metabolite, human gut microbiota metabolites, microorganisms.
Catalog No. 50-004-08560 Supplier Medchemexpress LLC Supplier No. HYB0671R50MG
Vancomycin (Standard) is the analytical standard of Vancomycin, intended for research and analytical applications. It is an antibiotic used for the treatment of bacterial infections.
  • The compound is the grade of analytical standard, which is the reference standard supplied assay.
  • It is commonly used in qualitative, quantitative, and methodological research experiments in HPLC, GC, and MS.
  • Application in disease research: Antibacterial.
  • Initial source: Endogenous metabolite, human gut microbiota metabolites, microorganisms.