Learn More
Medchemexpress LLC HY-10260 10mg , Vandetanib CAS:443913-73-3 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1026010MG
Encompass_Preferred
HY-10260 10mg Vandetanib CAS: 443913-73-3 Vandetanib (D6474) is a potent, orally active inhibitor of VEGFR2/KDR tyrosine kinase activity (IC50=40 nM). Vandetanib also has activity versus the tyrosine kinase activity of VEGFR3/FLT4 (IC50=110 nM) and EGFR/HER1 (IC50=500 nM). Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-225-9900