Learn More
MyBioSource VANGL2 antibody
SDP
Supplier:  MyBioSource MBS1570710
Encompass_Preferred
The VANGL2 antibody from MyBioSource is a rabbit polyclonal antibody raised against a recombinant protein of human VANGL2. This antibody detects endogenous levels of total VANGL2 protein. It can be used in Western blot, ELISA, and immunohistochemistry. The immunogen is a recombinant protein of human VANGL2. This antibody has been tested in human, mouse, and rat samples. The recommended dilution for Western blot is 1:500-1:1000, for ELISA it is 1:500-1:1000, and for IHC it is 1:50-1:200. This antibody is stored in PBS with 0.02% sodium azide and 50% glycerol.
Catalog No. NC2906308