Learn More
Waters Corp Waters Corporation Cartridge Holder, 1/pk, Ultra-Low Dispersion, 18,000 psi
Catalog No. 501093118 Shop All Waters Corporation Products
Encompass_Preferred
The VanGuard Cartridge Holder offers an ultra-low dispersion design with Fully Integrated Technology (FIT) for reliable connections, optimizing performance in critical separations by extending column lifetime and preventing contamination.
  • Ultra-low dispersion format for enhanced performance.
  • Features Fully Integrated Technology (FIT) for sound connections and reduced extra-column volume.
  • Operates at pressures up to 18,000 psi (1240 bar).
  • Protects UPLC, UHPLC, and HPLC columns (5 to 16 μm particle sizes).
  • Extends column lifespan by removing particulate and chemical contamination.
  • Ensures consistent, reproducible results with low dead volume design.
  • Two-piece design with a reusable holder and a replaceable cartridge.
Catalog No. 50-109-3118 Supplier Waters Corp Supplier No. 186007949
The VanGuard Cartridge Holder offers an ultra-low dispersion design with Fully Integrated Technology (FIT) for reliable connections, optimizing performance in critical separations by extending column lifetime and preventing contamination.
  • Ultra-low dispersion format for enhanced performance.
  • Features Fully Integrated Technology (FIT) for sound connections and reduced extra-column volume.
  • Operates at pressures up to 18,000 psi (1240 bar).
  • Protects UPLC, UHPLC, and HPLC columns (5 to 16 μm particle sizes).
  • Extends column lifespan by removing particulate and chemical contamination.
  • Ensures consistent, reproducible results with low dead volume design.
  • Two-piece design with a reusable holder and a replaceable cartridge.