Learn More
Med Vet International ZoetisVanguard Plus 5 Vaccine, Modified Live Virus, 25 X 1mL
Catalog No. 502188533
Encompass_Preferred
Canine Distemper-Adenovirus Type 2-Parainfluenza-Parvovirus Vaccine, Modified Live Virus, dogs 6 weeks or older. Preservative: Gentamicin.Shipping charges for refrigerated items are a flat charge based on your location or zone. Cold items are packed on ice and shipped separately in a cooler with expedited shipping. Med-Vet ships cold items Monday-Wednesday only. Next day air available for additional charge.
Catalog No. 50-218-8533 Supplier Med Vet International Supplier No. PHVVANGUARDP5
Canine Distemper-Adenovirus Type 2-Parainfluenza-Parvovirus Vaccine, Modified Live Virus, dogs 6 weeks or older. Preservative: Gentamicin.Shipping charges for refrigerated items are a flat charge based on your location or zone. Cold items are packed on ice and shipped separately in a cooler with expedited shipping. Med-Vet ships cold items Monday-Wednesday only. Next day air available for additional charge.