Learn More
Medchemexpress LLC Vanillic acid | 121-34-6 | 168.15 | 1 G
SDP
Catalog No. 5000417110
Encompass_Preferred
Vanillic acid is a flavoring agent found in edible plants and fruits, including Angelica sinensis. It acts as an inhibitor of NF-κB activation and exhibits anti-inflammatory, antibacterial, and chemopreventive effects. This product is intended for research use only.
  • Acts as an inhibitor of NF-κB activation
  • Exhibits anti-inflammatory, antibacterial, and chemopreventive effects
  • Found in various edible plants and fruits
  • Considered an endogenous metabolite
Catalog No. 50-004-17110 Supplier Medchemexpress LLC Supplier No. HYN07081G
Vanillic acid is a flavoring agent found in edible plants and fruits, including Angelica sinensis. It acts as an inhibitor of NF-κB activation and exhibits anti-inflammatory, antibacterial, and chemopreventive effects. This product is intended for research use only.
  • Acts as an inhibitor of NF-κB activation
  • Exhibits anti-inflammatory, antibacterial, and chemopreventive effects
  • Found in various edible plants and fruits
  • Considered an endogenous metabolite