Learn More
Medchemexpress LLC Vanillic acid | 121-34-6 | 168.15 | 1 ML
SDP
Catalog No. 5000415887
Encompass_Preferred
Vanillic acid is a flavoring agent found in edible plants and fruits, including Angelica sinensis. It functions by inhibiting NF-κB activation, and exhibits anti-inflammatory, antibacterial, and chemopreventive effects.
  • Flavoring agent
  • Found in edible plants and fruits, such as Angelica sinensis
  • Inhibits NF-κB activation
  • Anti-inflammatory
  • Antibacterial
  • Chemopreventive effects
Catalog No. 50-004-15887 Supplier Medchemexpress LLC Supplier No. HYN070810MM/1ML
Vanillic acid is a flavoring agent found in edible plants and fruits, including Angelica sinensis. It functions by inhibiting NF-κB activation, and exhibits anti-inflammatory, antibacterial, and chemopreventive effects.
  • Flavoring agent
  • Found in edible plants and fruits, such as Angelica sinensis
  • Inhibits NF-κB activation
  • Anti-inflammatory
  • Antibacterial
  • Chemopreventive effects