Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Vanillic acid | 121-34-6 | 168.15 | 100 MG
SDP

Catalog No. 5000416699
Encompass_Preferred

Vanillic acid is a flavoring agent present in edible plants and fruits, including Angelica sinensis. It acts by inhibiting NF-κB activation.

  • Found in edible plants, fruits, and Angelica sinensis.
  • Inhibits NF-κB activation.
  • Displays anti-inflammatory, antibacterial, and chemopreventive properties.
  • Purity: 99.43%.
  • Intended for research purposes only.
  • Soluble in DMSO (≥ 100 mg/mL).
  • Shows no toxicity to HT22 cells at concentrations of 50, 100, and 200 μM.
  • Significantly increases cell viability when co-treated with Aβ1-42.
  • Inhibits Carrageenan-induced mechanical hyperalgesia, paw edema, and immune cell recruitment in mice.
  • Appearance: Off-white to light brown solid.
  • Storage: Stable at room temperature for 3 years (solid); -80°C for 2 years or -20°C for 1 year (in solvent).

Catalog No. 50-004-16699 Supplier Medchemexpress LLC Supplier No. HYN0708100MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Vanillic acid is a flavoring agent present in edible plants and fruits, including Angelica sinensis. It acts by inhibiting NF-κB activation.

  • Found in edible plants, fruits, and Angelica sinensis.
  • Inhibits NF-κB activation.
  • Displays anti-inflammatory, antibacterial, and chemopreventive properties.
  • Purity: 99.43%.
  • Intended for research purposes only.
  • Soluble in DMSO (≥ 100 mg/mL).
  • Shows no toxicity to HT22 cells at concentrations of 50, 100, and 200 μM.
  • Significantly increases cell viability when co-treated with Aβ1-42.
  • Inhibits Carrageenan-induced mechanical hyperalgesia, paw edema, and immune cell recruitment in mice.
  • Appearance: Off-white to light brown solid.
  • Storage: Stable at room temperature for 3 years (solid); -80°C for 2 years or -20°C for 1 year (in solvent).

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.