Learn More
Medchemexpress LLC Vanillic acid | 121-34-6 | 168.15 | 500 MG
SDP
Catalog No. 5000415467
Encompass_Preferred
Vanillic acid is a flavoring agent found in edible plants and fruits, including Angelica sinensis. It functions by inhibiting NF-κB activation, and exhibits anti-inflammatory, antibacterial, and chemopreventive effects. It is intended for research use only and not for sale to patients.
  • Purity: 99.43% (Initial) and 99.75% (Specific Batch)
  • Inhibits NF-κB activation.
  • Anti-inflammatory effects.
  • Antibacterial effects.
  • Chemopreventive effects.
  • Non-toxic to HT22 cells at 50, 100, and 200 μM, and significantly increases cell viability when co-treated with Aβ1-42.
  • Inhibits Carrageenan-induced mechanical hyperalgesia, paw edema, and neutrophil and macrophage recruitment in mice at doses of 3-30 mg/kg.
Catalog No. 50-004-15467 Supplier Medchemexpress LLC Supplier No. HYN0708500MG
Vanillic acid is a flavoring agent found in edible plants and fruits, including Angelica sinensis. It functions by inhibiting NF-κB activation, and exhibits anti-inflammatory, antibacterial, and chemopreventive effects. It is intended for research use only and not for sale to patients.
  • Purity: 99.43% (Initial) and 99.75% (Specific Batch)
  • Inhibits NF-κB activation.
  • Anti-inflammatory effects.
  • Antibacterial effects.
  • Chemopreventive effects.
  • Non-toxic to HT22 cells at 50, 100, and 200 μM, and significantly increases cell viability when co-treated with Aβ1-42.
  • Inhibits Carrageenan-induced mechanical hyperalgesia, paw edema, and neutrophil and macrophage recruitment in mice at doses of 3-30 mg/kg.