Learn More
Apexbio Technology LLC Vanillic acid 121-34-6 500mg
SDP
Catalog No. 5000091159
Encompass_Preferred
Vanillic acid (121-34-6) is a small-molecule inhibitor modulating inflammatory pathways potentially via inhibition of pro-inflammatory cytokines and NF- B signaling It is designed to reduce intracellular oxidative stress and regulate inflammatory and antimicrobial processes Vanillic acid exerts its biological activity primarily through antioxidant and anti-inflammatory mechanisms of action In cellular and biochemical assays vanillic acid demonstrates inhibitory activity against selected microbial strains with IC50 values within the micromolar to millimolar range depending on assay conditions and organism types Based on these pharmacological properties vanillic acid holds research potential in antimicrobial and inflammation-related application fields
Catalog No. 50-000-91159 Supplier Apexbio Technology LLC Supplier No. B7917500
Vanillic acid (121-34-6) is a small-molecule inhibitor modulating inflammatory pathways potentially via inhibition of pro-inflammatory cytokines and NF- B signaling It is designed to reduce intracellular oxidative stress and regulate inflammatory and antimicrobial processes Vanillic acid exerts its biological activity primarily through antioxidant and anti-inflammatory mechanisms of action In cellular and biochemical assays vanillic acid demonstrates inhibitory activity against selected microbial strains with IC50 values within the micromolar to millimolar range depending on assay conditions and organism types Based on these pharmacological properties vanillic acid holds research potential in antimicrobial and inflammation-related application fields