Learn More
Medchemexpress LLC Vanillic acid | 121-34-6 | MFCD00002551 | 168.15 | 100 MG
SDP
Catalog No. 5000416663
Encompass_Preferred
Vanillic acid (Standard) is the analytical standard of Vanillic acid. This product is intended for research and analytical applications. It is a flavoring agent found in edible plants and fruits, including Angelica sinensis.
  • Inhibits NF-κB activation.
  • Has anti-inflammatory effects.
  • Has antibacterial effects.
  • Has chemopreventive effects.
Catalog No. 50-004-16663 Supplier Medchemexpress LLC Supplier No. HYN0708R100MG
Vanillic acid (Standard) is the analytical standard of Vanillic acid. This product is intended for research and analytical applications. It is a flavoring agent found in edible plants and fruits, including Angelica sinensis.
  • Inhibits NF-κB activation.
  • Has anti-inflammatory effects.
  • Has antibacterial effects.
  • Has chemopreventive effects.