Learn More
Selleck Chemical LLC Vanillic acid S5343-1g
SDP
Catalog No. 503109966
Encompass_Preferred
Vanillic acid (4-hydroxy-3-methoxybenzoic acid) is a flavoring agent which is also an intermediate in the production of vanillin from ferulic acid Vanillic acid inhibits NF- B activation Anti-inflammatory antibacterial and chemopreventive effects
Catalog No. 50-310-9966 Supplier Selleck Chemical LLC Supplier No. S53431G
Vanillic acid (4-hydroxy-3-methoxybenzoic acid) is a flavoring agent which is also an intermediate in the production of vanillin from ferulic acid Vanillic acid inhibits NF- B activation Anti-inflammatory antibacterial and chemopreventive effects