Learn More
Selleck Chemical LLC Vanillic acid S5343-25mg
SDP
Catalog No. 503110323
Encompass_Preferred
Vanillic acid (4-hydroxy-3-methoxybenzoic acid) is a flavoring agent which is also an intermediate in the production of vanillin from ferulic acid Vanillic acid inhibits NF- B activation Anti-inflammatory antibacterial and chemopreventive effects
Catalog No. 50-311-0323 Supplier Selleck Chemical LLC Supplier No. S534325MG
Vanillic acid (4-hydroxy-3-methoxybenzoic acid) is a flavoring agent which is also an intermediate in the production of vanillin from ferulic acid Vanillic acid inhibits NF- B activation Anti-inflammatory antibacterial and chemopreventive effects