Learn More
Medchemexpress LLC Vanillic acid glucos 10mg | 32142-31-7 | 10 MG
SDP
Catalog No. 5000229699
Encompass_Preferred
Vanillic acid glucoside is the 4-β-D-glucoside derivative of vanillic acid (CAS 32142-31-7), provided as a research-grade analytical standard (molecular formula C14H18O9; molecular weight 330.29 g/mol) typically in 10 mg vials for laboratory and analytical use.
  • High-purity analytical standard (~98.2%).
  • Well-characterized: molecular weight 330.29 g/mol, formula C14H18O9.
  • Suitable for analytical, biochemical, and natural product research.
  • Supplied as a milligram-scale vial for small-scale experiments.
Catalog No. 50-002-29699 Supplier Medchemexpress LLC Supplier No. HY11476010MG
Vanillic acid glucoside is the 4-β-D-glucoside derivative of vanillic acid (CAS 32142-31-7), provided as a research-grade analytical standard (molecular formula C14H18O9; molecular weight 330.29 g/mol) typically in 10 mg vials for laboratory and analytical use.
  • High-purity analytical standard (~98.2%).
  • Well-characterized: molecular weight 330.29 g/mol, formula C14H18O9.
  • Suitable for analytical, biochemical, and natural product research.
  • Supplied as a milligram-scale vial for small-scale experiments.