Learn More
Medchemexpress LLC Vanillic acid (ring-13C6) | 201612-56-8 | MFCD00145520 | 99.9% | 174.10 | C213C6H8O4 | 5mg
SDP
Catalog No. 5000268494
Encompass_Preferred
Vanillic acid-13C6 is the 13C labeled isotope of Vanillic acid-13C6(HY-N0708 ) Vanillic acid is a flavoring agent found in edible plants and fruits and in the root of angelica Vanillic acid inhibits NF- B activation It has anti-inflammatory and antimicrobial activities
Catalog No. 50-002-68494 Supplier Medchemexpress LLC Supplier No. HYW7782255MG
Vanillic acid-13C6 is the 13C labeled isotope of Vanillic acid-13C6(HY-N0708 ) Vanillic acid is a flavoring agent found in edible plants and fruits and in the root of angelica Vanillic acid inhibits NF- B activation It has anti-inflammatory and antimicrobial activities