Learn More
Medchemexpress LLC Vanillic acid (Standard) | 121-34-6 | 168.15 | 50 MG
SDP
Catalog No. 5000416664
Encompass_Preferred
Vanillic acid (Standard) is the analytical standard of Vanillic acid, intended for research and analytical applications. It is a flavoring agent found in edible plants and fruits, including *Angelica sinensis*. This compound inhibits NF-κB activation and exhibits anti-inflammatory, antibacterial, and chemopreventive effects. It is commonly used in qualitative, quantitative, and methodological research experiments in HPLC, GC, and MS.
  • Analytical standard grade
  • Reference standard supplied assay
  • Inhibits NF-κB activation
  • Exhibits anti-inflammatory effects
  • Exhibits antibacterial effects
  • Exhibits chemopreventive effects
Catalog No. 50-004-16664 Supplier Medchemexpress LLC Supplier No. HYN0708R50MG
Vanillic acid (Standard) is the analytical standard of Vanillic acid, intended for research and analytical applications. It is a flavoring agent found in edible plants and fruits, including *Angelica sinensis*. This compound inhibits NF-κB activation and exhibits anti-inflammatory, antibacterial, and chemopreventive effects. It is commonly used in qualitative, quantitative, and methodological research experiments in HPLC, GC, and MS.
  • Analytical standard grade
  • Reference standard supplied assay
  • Inhibits NF-κB activation
  • Exhibits anti-inflammatory effects
  • Exhibits antibacterial effects
  • Exhibits chemopreventive effects