Learn More
Medchemexpress LLC Vanillin-13C6 | 201595-58-6 | ≥99.0% | 158.10 | 5 MG
SDP
Catalog No. 5000427694
Encompass_Preferred
Vanillin-13C6 is the 13C labeled Vanillin. Vanillin is a single molecule extracted from vanilla beans and is a popular odor used widely in perfume, food and medicine. Stable heavy isotopes of hydrogen, carbon, and other elements have been incorporated into drug molecules, largely as tracers for quantitation during the drug development process. Deuteration has gained attention because of its potential to affect the pharmacokinetic and metabolic profiles of drugs.
  • Can be used as a tracer
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
Catalog No. 50-004-27694 Supplier Medchemexpress LLC Supplier No. HYN0098S25MG
Vanillin-13C6 is the 13C labeled Vanillin. Vanillin is a single molecule extracted from vanilla beans and is a popular odor used widely in perfume, food and medicine. Stable heavy isotopes of hydrogen, carbon, and other elements have been incorporated into drug molecules, largely as tracers for quantitation during the drug development process. Deuteration has gained attention because of its potential to affect the pharmacokinetic and metabolic profiles of drugs.
  • Can be used as a tracer
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS