Learn More
Apexbio Technology LLC Vanillin 121-33-5 200mg
SDP
Catalog No. 502240548
Encompass_Preferred
Vanillin (CAS 121-33-5) is an aromatic aldehyde recognized as the principal constituent of vanilla bean extract Beyond its traditional use as a flavoring agent vanillin exhibits diverse biological activities including antioxidant antiangiogenic and anti-inflammatory effects Mechanistic studies suggest vanillin modulates hemoglobin polymerization contributing to its anti-sickling properties relevant to sickle cell anemia research It has also demonstrated attenuation of ultraviolet A-induced loss of stemness in human adipose-derived mesenchymal stem cells and exhibits antidepressant-like effects by influencing monoamine neurotransmitter levels in the brain Vanillin is thus under investigation for therapeutic roles in hematologic neurological and regenerative medicine contexts
This item is not returnable. View return policy
Catalog No. 50-224-0548 Supplier Apexbio Technology LLC Supplier No. B3679200
Vanillin (CAS 121-33-5) is an aromatic aldehyde recognized as the principal constituent of vanilla bean extract Beyond its traditional use as a flavoring agent vanillin exhibits diverse biological activities including antioxidant antiangiogenic and anti-inflammatory effects Mechanistic studies suggest vanillin modulates hemoglobin polymerization contributing to its anti-sickling properties relevant to sickle cell anemia research It has also demonstrated attenuation of ultraviolet A-induced loss of stemness in human adipose-derived mesenchymal stem cells and exhibits antidepressant-like effects by influencing monoamine neurotransmitter levels in the brain Vanillin is thus under investigation for therapeutic roles in hematologic neurological and regenerative medicine contexts