Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Vanillin acetate | 881-68-5 | 99.94% | 194.19 | 25 G
SDP

Catalog No. 5000452405
Encompass_Preferred

Vanillin acetate is a phenol ester that can be synthesized from vanillin by treatment with acetic anhydride. It contains a benzene ring substituted by a hydroxyl group and an ester group. It is intended for research use only.

  • Phenol ester.
  • Synthesized from vanillin by treatment with acetic anhydride.
  • Contains a benzene ring substituted by a hydroxyl group and an ester group.
  • Available in various quantities.
  • Comes with a data sheet, COA, SDS, and handling instructions.
  • Soluble in DMSO (≥ 100 mg/mL).
  • Can be prepared in stock solutions with various concentrations.
  • Protocols for in vivo dissolution are provided using combinations of DMSO, PEG300, Tween-80, Saline, SBE-β-CD, and Corn Oil.
  • Storage conditions: 4°C (stored under nitrogen) for the product itself; -80°C (6 months) or -20°C (1 month, stored under nitrogen) for solutions.

Catalog No. 50-004-52405 Supplier Medchemexpress LLC Supplier No. 25GHYW009948
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Vanillin acetate is a phenol ester that can be synthesized from vanillin by treatment with acetic anhydride. It contains a benzene ring substituted by a hydroxyl group and an ester group. It is intended for research use only.

  • Phenol ester.
  • Synthesized from vanillin by treatment with acetic anhydride.
  • Contains a benzene ring substituted by a hydroxyl group and an ester group.
  • Available in various quantities.
  • Comes with a data sheet, COA, SDS, and handling instructions.
  • Soluble in DMSO (≥ 100 mg/mL).
  • Can be prepared in stock solutions with various concentrations.
  • Protocols for in vivo dissolution are provided using combinations of DMSO, PEG300, Tween-80, Saline, SBE-β-CD, and Corn Oil.
  • Storage conditions: 4°C (stored under nitrogen) for the product itself; -80°C (6 months) or -20°C (1 month, stored under nitrogen) for solutions.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.