Learn More
U.S. Pharmacopeia Vanillin Melting Point Standard, 121-33-5, MFCD00006942, 1 g
Catalog No. NC9849081
Encompass_Preferred
Linear Formula: 4-(HO)C6H3-3-(OCH3)CHO, Molecular Weight: 152.15, BP: 170 °C/15 mmHg, MP: 81-83 °C, Synonym: 4-Hydroxy-3-methoxybenzaldehyde, Vanillic aldehyde, Vanillin.
Catalog No. NC9849081 Supplier U.S. Pharmacopeia Supplier No. 1711009
Linear Formula: 4-(HO)C6H3-3-(OCH3)CHO, Molecular Weight: 152.15, BP: 170 °C/15 mmHg, MP: 81-83 °C, Synonym: 4-Hydroxy-3-methoxybenzaldehyde, Vanillic aldehyde, Vanillin.