Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Vanillin, NF, 97-103%, Spectrum™ Chemical
SDP

Catalog No. 18607698 Shop All Spectrum Chemical Manufacturing Corporation Products
Change view
Click to view available options
Quantity:
125 g
500 g
2.5 kg
12 kg
50 kg
Packaging:
Amber Glass Bottle
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18-607-698 500 g Amber Glass Bottle
18-607-697 125 g Amber Glass Bottle
18-607-699 2.5 kg Amber Glass Bottle
18-607-700 12 kg Amber Glass Bottle
18-607-701 50 kg Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 18-607-698 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. VA130500GM
Add to Cart
Add to Cart

VA130, 121-33-5, C8H8O3

Spectrum™ Chemical Vanillin, NF is used as a flavoring agent in pharmaceutical formulations. All Spectrum Chemical manufactured NF grade products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Chemical Identifiers

CAS 121-33-5
Molecular Formula C8H8O3
Molecular Weight (g/mol) 152.15
MDL Number MFCD00006942,MFCD08702848
InChI Key MWOOGOJBHIARFG-UHFFFAOYSA-N
IUPAC Name 4-hydroxy-3-methoxybenzaldehyde
SMILES COC1=CC(C=O)=CC=C1O

Specifications

CAS Max % 1
Ignition Residue 0.0005
Loss on Drying 0.01
Melting Range 81-83°C
Packaging Amber Glass Bottle
Quantity 500 g
Percent Purity 97 to 103%
Grade NF
Physical Form Crystals or Flakes
Chemical Name or Material Vanillin
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.