Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Vanillin OR, Macron™

Catalog No. MK2759500 Shop All Avantor Products
Change view
Click to view available options
Glass; 500g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
MK-2759-500 Glass; 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MK-2759-500 Supplier Avantor Supplier No. 275959

missing translation for 'validMainframeMsg'

Specifications

CAS 121-33-5
Molecular Formula C8H8O3
MDL Number MFCD00006942,MFCD08702848
Packaging Glass
Quantity 500 g
Synonym vanillin, vanillaldehyde, vanillic aldehyde, vanilla, p-vanillin, 2-methoxy-4-formylphenol, vanilline, lioxin, 4-hydroxy-m-anisaldehyde, 3-methoxy-4-hydroxybenzaldehyde
InChI Key MWOOGOJBHIARFG-UHFFFAOYSA-N
SMILES COC1=CC(C=O)=CC=C1O
IUPAC Name 4-hydroxy-3-methoxybenzaldehyde
Molecular Weight (g/mol) 152.15
PubChem CID 1183
ChEBI CHEBI:18346
Formula Weight 152.15
Chemical Name or Material Vanillin
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.