Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Vanillin Pharmaceutical Secondary Standard, MilliporeSigma™ Supelco™

Catalog No. 111012768 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-101-2768 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-101-2768 Supplier MilliporeSigma Supelco Supplier No. PHR12451G
Add to Cart
Add to Cart

Vanillin is an aromatic compound, and is used for flavoring foods. It is has biogenetic relationship to phenylpropanoid pathway. It is mostly used in foods, beverages, perfumes and pharmaceuticals.

Cleaning Products, Cosmetics, Food and Beverages, Personal Care and Pharmaceutical (small molecule)
TRUSTED_SUSTAINABILITY

Specifications

CAS 121-33-5
Vapor Pressure >0.01 mmHg (25°C)
Vapor Density 5.3 (vs. air)
Flash Point 153°C - Closed Cup
Physical Form Neat
Linear Formula 4-(HO)C6H3-3-(OCH3)CHO
MDL Number MFCD00006942
Melting Point 81°C to 83°C
Boiling Point 170°C (15 mmHg)
Synonym 4-Hydroxy-3-methoxybenzaldehyde; Vanillic aldehyde
Type Certified Reference Material
Quantity 1 g
Formula Weight 152.15
Grade Certified Reference Material
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.