Learn More
Novus Biologicals™ Vanilloid R1/TRPV1 Peptide

Specifications
Specifications
| Gene ID (Entrez) | 7442 |
| Species | Rat |
| Content And Storage | Store at -80°C. Avoid freeze-thaw cycles. |
| For Use With (Application) | This peptide is useful as a blocking peptide for NBP1-97417. For further blocking peptide related protocol, click here . |
| Gene Alias | Capsaicin receptor, Osm-9-like TRP channel 1, OTRPC1, transient receptor potential cation channel subfamily V member 1, transient receptor potential cation channel, subfamily V, member 1, transient receptor potential vanilloid 1a, transient receptor potential vanilloid 1b, TrpV1, Vanilloid receptor 1, vanilloid receptor subtype 1, VR1, VR1DKFZp434K0220 |
| Gene Symbol | TRPV1 |
| Quantity | 0.05 mg |
| Research Category | Neuroendocrinology, Neuroscience, Sensory Systems |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.