Learn More
Selleck Chemical LLC Vanillyl Butyl Ether S3965-25mg
SDP
Catalog No. 503108804
Encompass_Preferred
Vanillyl Butyl Ether (4-(Butoxymethyl)-2-methoxyphenol) an ether of monohydroxybenzoic acid is added to food products as a flavoring agent It is also present in cosmetics and personal care products as a fragrance ingredient oral care agent hair conditioning agent and warming or cooling agent
Catalog No. 50-310-8804 Supplier Selleck Chemical LLC Supplier No. S396525MG
Vanillyl Butyl Ether (4-(Butoxymethyl)-2-methoxyphenol) an ether of monohydroxybenzoic acid is added to food products as a flavoring agent It is also present in cosmetics and personal care products as a fragrance ingredient oral care agent hair conditioning agent and warming or cooling agent