Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Vanillyl butyl ether | 82654-98-6 | MFCD00238529 | 99.7% | 210.27 g/mol | C12H18O3 | 100 G
SDP

Catalog No. 5000303915
Encompass_Preferred

Vanillyl butyl ether (CAS 82654-98-6) is an organic research reagent with warming and insect-repellent activity, used in biological screening, formulation, and fragrance studies. It is supplied as a colorless to light yellow liquid and is available as a DMSO solution for assay work.

  • Molecular formula C12H18O3 and molecular weight 210.27 g/mol.
  • Assay 99.7% by GC, water 0.85% (batch COA).
  • Appearance: colorless to light yellow liquid.
  • Available as 10 mM solution in DMSO, and in 25 g, 50 g, 100 g quantities.
  • Recommended storage: pure form -20°C (3 years) or 4°C (2 years); in solvent -80°C (6 months).
  • Suitable for repellent, warming, and sensory research applications.

Catalog No. 50-003-03915 Supplier Medchemexpress LLC Supplier No. HYW014394100G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Vanillyl butyl ether (CAS 82654-98-6) is an organic research reagent with warming and insect-repellent activity, used in biological screening, formulation, and fragrance studies. It is supplied as a colorless to light yellow liquid and is available as a DMSO solution for assay work.

  • Molecular formula C12H18O3 and molecular weight 210.27 g/mol.
  • Assay 99.7% by GC, water 0.85% (batch COA).
  • Appearance: colorless to light yellow liquid.
  • Available as 10 mM solution in DMSO, and in 25 g, 50 g, 100 g quantities.
  • Recommended storage: pure form -20°C (3 years) or 4°C (2 years); in solvent -80°C (6 months).
  • Suitable for repellent, warming, and sensory research applications.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.