Learn More
Medchemexpress LLC Vanillylmandelic acid | 55-10-7 | 99.62% | 198.17 | 100 MG
SDP
Supplier:  Medchemexpress LLC HY113121R100MG
Encompass_Preferred
Vanillylmandelic acid (Standard) is an analytical standard intended for research and analytical applications. It is the end product of epinephrine and norepinephrine metabolism and can be used as an indication of disorders in neurotransmitter metabolism. It also exhibits antioxidant activity towards DPPH radical with an IC50 value of 33 μM.
  • Used as a reference standard for assay
  • Utilized in qualitative, quantitative, and methodological research experiments in HPLC, GC, and MS
Catalog No. 50-004-35949