Learn More
Roboz Surgical VANNAS 3X.2MM STR
SDP
Catalog No. NC9457428
Encompass
Vannas Micro Dissecting Spring Scissors; Straight; 3mm Cutting Edge; 0.1mm Tip Width; 3 in. Overall Length These scissors have finer blades than the very similar RS-5610 scissors. The tips are narrower and the characteristic notch in the blades of this
Catalog No. NC9457428 Supplier Roboz Surgical Supplier No. RS5620
Vannas Micro Dissecting Spring Scissors; Straight; 3mm Cutting Edge; 0.1mm Tip Width; 3 in. Overall Length These scissors have finer blades than the very similar RS-5610 scissors. The tips are narrower and the characteristic notch in the blades of this