Learn More
Roboz Surgical VANNAS ANGLE ON EDGE
SDP
Catalog No. NC9508001
Encompass
Vannas Micro Dissecting Spring Scissors; Angled On Edge (Points To Left When Laid Down); 3mm Cutting Edge; 0.15mm Tip Width; 3 in. Overall Length Angled blades make it easier to cut from above while keeping the blades aligned with the line of cutting s
Catalog No. NC9508001 Supplier Roboz Surgical Supplier No. RS5618
Vannas Micro Dissecting Spring Scissors; Angled On Edge (Points To Left When Laid Down); 3mm Cutting Edge; 0.15mm Tip Width; 3 in. Overall Length Angled blades make it easier to cut from above while keeping the blades aligned with the line of cutting s