Learn More
World Precision Instrument Vannas Capsulatomy Scissors, 9.5cm (3.75), Curved Sharp Tips, Non-Magnetic, Titanium
SDP
Catalog No. 502538290
Encompass
  • 9.5cm (3.7") Long
  • Cutting edge 7.5 mm
  • Sharp tips
  • Titanium
Catalog No. 50-253-8290 Supplier World Precision Instrument Supplier No. WP5060
  • 9.5cm (3.7") Long
  • Cutting edge 7.5 mm
  • Sharp tips
  • Titanium