Learn More
Roboz Surgical Roboz Surgical Vannas Spring Scissors, Curved, 3.5in Length, 1 ea
SDP
Catalog No. NC9186188
Encompass
These micro dissecting spring scissors feature a precise curved design for delicate surgical procedures. Engineered for fine-scale tissue manipulation, these instruments provide exceptional control and accuracy in microsurgical environments.
  • Curved geometry to facilitate access in confined surgical sites
  • Fine blades with a 3 mm cutting edge for clean, precise incisions
  • Integrated spring mechanism for smooth, responsive operation
  • Durable stainless steel construction suitable for standard sterilization protocols
Catalog No. NC9186188 Supplier Roboz Surgical Supplier No. RS5611
These micro dissecting spring scissors feature a precise curved design for delicate surgical procedures. Engineered for fine-scale tissue manipulation, these instruments provide exceptional control and accuracy in microsurgical environments.
  • Curved geometry to facilitate access in confined surgical sites
  • Fine blades with a 3 mm cutting edge for clean, precise incisions
  • Integrated spring mechanism for smooth, responsive operation
  • Durable stainless steel construction suitable for standard sterilization protocols