Learn More
Roboz Surgical VANNAS MICRO DISSECTING SCISSO
SDP
Catalog No. NC1625543
Encompass
Vannas Micro Dissecting Spring Scissors. Angled On Flat Angles Up When Laid Down. 3mm Cutting.Edge; 0.15mm Tip Width; 3" Overall Length
Catalog No. NC1625543 Supplier Roboz Surgical Supplier No. RS5619
Vannas Micro Dissecting Spring Scissors. Angled On Flat Angles Up When Laid Down. 3mm Cutting.Edge; 0.15mm Tip Width; 3" Overall Length