Learn More
World Precision Instrument WPI Premium Vannas Spring Scissors, 9 cm Long, 3 mm Tips, 6 mm Blades
SDP
Catalog No. 50379790
Encompass
Our Vannas scissors are specialized spring scissors designed for precision cutting and often used by scientific researchers. They are named after the Swedish scientist Gunnar Vannas who invented them in the 1940s. Compared with other spring scissors, Vannas scissors are distinguished by their delicate, thin, sharp tips. These surgical scissors are 9cm long and have 6mm straight blades and sharp tips. The slender, pointed tips allows for precise cutting with minimal damage to surrounding tissues. Choose between straight or angled on the flat blades. Like all spring scissors, these are an excellent choice for left-handed operators, because they can be used ambidextrously. These Vannas scissors are made of surgical grade stainless steel. To properly care for your surgical instruments, these scissors should be cleaned immediately after use to ensure a long service life and minimize staining or corrosion. These Vannas are autoclavable.
Catalog No. 50-379-790 Supplier World Precision Instrument Supplier No. 503371
Our Vannas scissors are specialized spring scissors designed for precision cutting and often used by scientific researchers. They are named after the Swedish scientist Gunnar Vannas who invented them in the 1940s. Compared with other spring scissors, Vannas scissors are distinguished by their delicate, thin, sharp tips. These surgical scissors are 9cm long and have 6mm straight blades and sharp tips. The slender, pointed tips allows for precise cutting with minimal damage to surrounding tissues. Choose between straight or angled on the flat blades. Like all spring scissors, these are an excellent choice for left-handed operators, because they can be used ambidextrously. These Vannas scissors are made of surgical grade stainless steel. To properly care for your surgical instruments, these scissors should be cleaned immediately after use to ensure a long service life and minimize staining or corrosion. These Vannas are autoclavable.