Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Electron Microscopy Sciences Vannas Scissors Angle On Side, Delicate, Extra fine
SDP

Catalog No. 5027489
Encompass_Preferred

All micro scissors have extra fine tips and are 3" (75 mm) long.
Cutting Edge: 5 mm
Tips: Extra fine

Catalog No. 50-274-89 Supplier Electron Microscopy Sciences Supplier No. 7293304
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

All micro scissors have extra fine tips and are 3" (75 mm) long.
Cutting Edge: 5 mm
Tips: Extra fine

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.