Learn More
Electron Microscopy Sciences MicroPoint™ Featherlite Vannas Scissors Straight
SDP
Catalog No. 5030627
Encompass_Preferred
MicroPoint Featherlite  Style DS-1 has straight Vannas Scissors has 5mm smooth blades.
OAL: 125 mm
Catalog No. 50-306-27 Supplier Electron Microscopy Sciences Supplier No. 77900S
MicroPoint Featherlite  Style DS-1 has straight Vannas Scissors has 5mm smooth blades.
OAL: 125 mm