Learn More
Fine Science Tools Vannas Spring Scissors, Microserrated, Straight, 8.5 cm
SDP
Catalog No. NC0901390
Encompass
Tip Shape: Straight/ Tips: Sharp/ Cutting Edge: 4mm/ Tip Diameter: 0.1mm/ Alloy / Material: Stainless Steel/ Length: 8.5cm/ Serrated: Yes/
Catalog No. NC0901390 Supplier Fine Science Tools Supplier No. 1507008
Tip Shape: Straight/ Tips: Sharp/ Cutting Edge: 4mm/ Tip Diameter: 0.1mm/ Alloy / Material: Stainless Steel/ Length: 8.5cm/ Serrated: Yes/