Learn More
Fine Science Tools VANNAS SPRING SCISSORS - TITAN
SDP
Catalog No. NC0526115
Encompass
Tip Shape: Straight/ Tips: Sharp/ Cutting Edge: 5mm/ Tip Diameter: 0.3mm/ Alloy / Material: Titanium/ Length: 9.5cm/ Serrated: No
Catalog No. NC0526115 Supplier Fine Science Tools Supplier No. 1561008
Tip Shape: Straight/ Tips: Sharp/ Cutting Edge: 5mm/ Tip Diameter: 0.3mm/ Alloy / Material: Titanium/ Length: 9.5cm/ Serrated: No