Learn More
World Precision Instrument Vannas Tubingen Scissors, 8cm, Curved, German Steel
SDP
Catalog No. 502539485
Encompass
Vannas Tübingen scissors are specialized microsurgical scissors that are highly precise and delicate, commonly used in ophthalmic and neurological procedures. They are named after Tübingen, a city in Germany, where the Max Planck Institute for Biological Cybernetics is located. Vannas Tübingen scissors are incredibly fine instruments, crafted of high-quality surgical grade stainless steel with extreme precision. Their design includes thin, and sharp blades, which allow for highly accurate and precise cutting in microsurgical settings. The tips of these scissors are exceptionally sharp and thin, enabling them to access and cut very fine and delicate tissues without causing unnecessary damage to surrounding structures. This is especially crucial in delicate surgeries, such as in ophthalmic procedures, where precision is paramount. The blades of Vannas Tübingen scissors can be curved or straight.
Catalog No. 50-253-9485 Supplier World Precision Instrument Supplier No. 503379
Vannas Tübingen scissors are specialized microsurgical scissors that are highly precise and delicate, commonly used in ophthalmic and neurological procedures. They are named after Tübingen, a city in Germany, where the Max Planck Institute for Biological Cybernetics is located. Vannas Tübingen scissors are incredibly fine instruments, crafted of high-quality surgical grade stainless steel with extreme precision. Their design includes thin, and sharp blades, which allow for highly accurate and precise cutting in microsurgical settings. The tips of these scissors are exceptionally sharp and thin, enabling them to access and cut very fine and delicate tissues without causing unnecessary damage to surrounding structures. This is especially crucial in delicate surgeries, such as in ophthalmic procedures, where precision is paramount. The blades of Vannas Tübingen scissors can be curved or straight.