Learn More
Fine Science Tools Vannas-Tübingen Spring Scissors
SDP
Catalog No. NC9321271
Encompass
Tip Shape: Curved/ Tips: Sharp/ Cutting Edge: 5mm/ Tip Diameter: 0.1mm/ Alloy / Material: Stainless Steel/ Length: 8.5cm/ Serrated: No
Catalog No. NC9321271 Supplier Fine Science Tools Supplier No. 1500408
Tip Shape: Curved/ Tips: Sharp/ Cutting Edge: 5mm/ Tip Diameter: 0.1mm/ Alloy / Material: Stainless Steel/ Length: 8.5cm/ Serrated: No