Learn More
Selleck Chemical LLC Vanoxerine dihydrochloride
SDP
Catalog No. 5000003336
Encompass_Preferred
Vanoxerine (GBR-12909) is a potent and selective inhibitor of the presynaptic dopamine uptake complex
Catalog No. 50-000-03336 Supplier Selleck Chemical LLC Supplier No. S579625MG
Vanoxerine (GBR-12909) is a potent and selective inhibitor of the presynaptic dopamine uptake complex