Learn More
Apexbio Technology LLC Vanoxerine dihydrochloride 67469-78-7 25mg
SDP
Catalog No. 5000094173
Encompass_Preferred
Vanoxerine dihydrochloride is a small-molecule inhibitor targeting the dopamine transporter (DAT1) human ether- -go-go-related gene (hERG Kv11 1) potassium channel cardiac sodium channel (hNaV1 5) and L-type calcium channels (ICa L) It modulates neurotransmitter uptake and cardiac ion channel conductance thereby regulating synaptic dopamine levels and cardiac electrophysiology Vanoxerine dihydrochloride exerts its biological activity primarily through inhibition of dopamine reuptake and blockade of multiple cardiac ion channels It demonstrates antagonist activity at DAT1 (Ki 16 9 nM) potent inhibition of hERG channels (IC50 0 84 nM) and inhibitory effects on hNaV1 5 (IC50 830 nM) and ICa L (IC50 320 nM) Based on these pharmacological properties vanoxerine dihydrochloride holds research potential in cardiac arrhythmia studies
Catalog No. 50-000-94173 Supplier Apexbio Technology LLC Supplier No. B324825
Vanoxerine dihydrochloride is a small-molecule inhibitor targeting the dopamine transporter (DAT1) human ether- -go-go-related gene (hERG Kv11 1) potassium channel cardiac sodium channel (hNaV1 5) and L-type calcium channels (ICa L) It modulates neurotransmitter uptake and cardiac ion channel conductance thereby regulating synaptic dopamine levels and cardiac electrophysiology Vanoxerine dihydrochloride exerts its biological activity primarily through inhibition of dopamine reuptake and blockade of multiple cardiac ion channels It demonstrates antagonist activity at DAT1 (Ki 16 9 nM) potent inhibition of hERG channels (IC50 0 84 nM) and inhibitory effects on hNaV1 5 (IC50 830 nM) and ICa L (IC50 320 nM) Based on these pharmacological properties vanoxerine dihydrochloride holds research potential in cardiac arrhythmia studies