Learn More
Thermo Scientific Deuterium Lamp 1 EA
Supplier:  Thermo Scientific 6083.1110
Encompass
Deuterium lamp light source with sealed arc tube containing deuterium gas that emits continuous 160NM to 400NM ultraviolet radiation spectrum when electrically excited, used as UV light source in spectroscopy instruments like UV-Vis spectrophotometers for quantitative chemical analysis, with 2000HR expected lifetime.
Catalog No. NC1312740