Learn More
Perkin Elmer US LLC Aluminum Vapor Pressure Pan Kit for TGA 8000 and Pyris 1 TGA
Catalog No. 509052213
Encompass_Preferred
The Aluminum Vapor Pressure Pan Kit is for use with DSC 4000, DSC 6000, DSC 8000, DSC 8500, Diamond DSC, Pyris 1 DSC, DSC 7, DSC 6, Pyris 6 DSC and Jade DSC. Requires Crimper/Sealer (02190061) and (B0139005 with B0144637).
Catalog No. 50-905-2213 Supplier Perkin Elmer US LLC Supplier No. N5190788
The Aluminum Vapor Pressure Pan Kit is for use with DSC 4000, DSC 6000, DSC 8000, DSC 8500, Diamond DSC, Pyris 1 DSC, DSC 7, DSC 6, Pyris 6 DSC and Jade DSC. Requires Crimper/Sealer (02190061) and (B0139005 with B0144637).