Learn More
Medchemexpress LLC Vapendavir diphospha 10mM 1mL | 1198151-75-5 | 10MM 1ML
SDP
Catalog No. 5000243742
Encompass_Preferred
Vapendavir diphosphate (BTA798 diphosphate) is a potent enteroviral capsid binder (CB) Vapendavir diphosphate (BTA798 diphosphate) possesses potent antiviral activity for enterovirus 71 (EV71) replication with EC50 values of 0 5-1 4 M in different EV71 strains[1 [2
Catalog No. 50-002-43742 Supplier Medchemexpress LLC Supplier No. HY106254A10MM 1ML
Vapendavir diphosphate (BTA798 diphosphate) is a potent enteroviral capsid binder (CB) Vapendavir diphosphate (BTA798 diphosphate) possesses potent antiviral activity for enterovirus 71 (EV71) replication with EC50 values of 0 5-1 4 M in different EV71 strains[1 [2