Learn More
DSI LLC VAPLOCK EF-100 VAPOR FLTR 5/PK
Catalog No. NC9497194
Encompass_Preferred
Vaplock EF-100V: Minimize odors by adsorption before they enter the lab environment. Constructed of activated coconut shell carbon & silica dessicant, with Polypropylene housing.
Catalog No. NC9497194 Supplier DSI LLC Supplier No. EF100V
Vaplock EF-100V: Minimize odors by adsorption before they enter the lab environment. Constructed of activated coconut shell carbon & silica dessicant, with Polypropylene housing.