Learn More
Cole-Parmer Spex VapLock™ Fitting, Polypropylene, Straight, Hose Barb to Threaded Adapter, 1/4" ID x 1/8" NPT(M); 1/EA
Catalog No. NC2119850 Shop All Cole-Parmer Products
Encompass_Preferred
Adapters for flexible tubing let you connect 1/16" to 3/4" ID soft-wall tubing to NPT(F) ports. Adapters are made of polypropylene. Wrap NPT threads 3 to 4 times with PTFE tape and wrench tighten for leak-resistant connections.
This item is not returnable. View return policy
Catalog No. NC2119850 Supplier Cole-Parmer Supplier No. 1202637
Adapters for flexible tubing let you connect 1/16" to 3/4" ID soft-wall tubing to NPT(F) ports. Adapters are made of polypropylene. Wrap NPT threads 3 to 4 times with PTFE tape and wrench tighten for leak-resistant connections.