Learn More
Cole-Parmer Port Plug, 1/4"-28 UNF(M), natural PTFE; 10/pk
Supplier:  Cole-Parmer 1202047
Encompass_Preferred
Cole-Parmer VapLock Port Plug, 1/4"-28 UNF(M), natural PTFE; 10/pk, Use plugs to seal unused ports for a leak-resistant systemSeal unused manifold and bottle cap ports
Catalog No. NC1413934
Add to Cart
Description
Description
Product Suggestions
Product Suggestions
Videos
Safety and Handling
Safety and Handling
WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov