Learn More
Cole-Parmer™ VapLock™ Solvent Waste Kits for Jars and Pails
Catalog No. 13311124
Change view
Click to view available options
Size:
100-415
70 mm Rieke
Material:
PTFE
Polypropylene
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size Material
13-311-124 70 mm Rieke Polypropylene
13-311-125 70 mm Rieke PTFE
13-311-148 100-415 Polypropylene
13-311-149 100-415 PTFE
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 13-311-124 Supplier Cole-Parmer™ Supplier No. 1201922
Add to Cart
Add to Cart
VapLock™ Solvent Waste Kits for Jars and Pails collect effluent from multiple sources while minimizing harmful solvent exhaust
Various cap sizes available to fit your solvent container
Warranty and Services
None
Specifications
For Use With (Equipment) Solvent-Compatible Containers
Material Polypropylene
Size 70 mm Rieke
Thread Size (English) 1/2 in. NPT(F), 1/4 in. NPT(F), 1/4 in.-28 UNF(F)
For Use With (Application) Solvent Safety
Type Solvent Waste Kit
WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov