Learn More
Cole-Parmer™ VapLock™ Solvent Waste Manifold Caps for Justrite™ Containers
Catalog No. 13311157
Change view
Click to view available options
Closure Size:
1/2 in. NPT(F), 1/4 in. NPT(F)
1/2 in. NPT(F), 1/4 in. NPT(F), 1/4 in.-28 UNF(F)
Material:
PTFE, Stainless Steel Fitting
Polypropylene, Polypropylene Fitting
Polypropylene, Stainless Steel Fitting
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Closure Size Material
13-311-157 1/2 in. NPT(F), 1/4 in. NPT(F), 1/4 in.-28 UNF(F) Polypropylene, Polypropylene Fitting
13-311-158 1/2 in. NPT(F), 1/4 in. NPT(F) Polypropylene, Polypropylene Fitting
13-311-159 1/2 in. NPT(F), 1/4 in. NPT(F), 1/4 in.-28 UNF(F) Polypropylene, Stainless Steel Fitting
13-311-160 1/2 in. NPT(F), 1/4 in. NPT(F) Polypropylene, Stainless Steel Fitting
13-311-161 1/2 in. NPT(F), 1/4 in. NPT(F), 1/4 in.-28 UNF(F) PTFE, Stainless Steel Fitting
13-311-162 1/2 in. NPT(F), 1/4 in. NPT(F) PTFE, Stainless Steel Fitting
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Catalog No. 13-311-157 Supplier Cole-Parmer™ Supplier No. 1201955
Add to Cart
Add to Cart
VapLock™ Solvent Waste Manifold Caps for Justrite™ Containers connect to waste containers for a secure closed system
Different materials, numbers and sizes of ports help customize to your application
Warranty and Services
None
Specifications
Closure Size 1/2 in. NPT(F), 1/4 in. NPT(F), 1/4 in.-28 UNF(F)
Closure Type Justrite Quick-Disconnect
For Use With (Application) Solvent Safety
For Use With (Equipment) Solvent-Compatible Containers
Material Polypropylene, Polypropylene Fitting
Product Type Solvent Waste Manifold