Learn More
Cole-Parmer VAPLOCK THREADED ADAPTER, PTF
Catalog No. NC1825844 Shop All Cole-Parmer Products
Encompass_Preferred
Cole-Parmer VapLock Threaded Adapter, PTFE, 1/4" NPT(M) x 1/4-28 UNF(F), 9/16" Hex; 1/EA
Catalog No. NC1825844 Supplier Cole-Parmer Supplier No. 1202082
Cole-Parmer VapLock Threaded Adapter, PTFE, 1/4" NPT(M) x 1/4-28 UNF(F), 9/16" Hex; 1/EA